missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-methylthiazole-5-carboxylate, 97%

Catalog No. AAA1689614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1689614 25 g
AAA1689606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1689614 Supplier Thermo Scientific Chemicals Supplier No. A1689614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7210-76-6 | C7H10N2O2S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7210-76-6
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00123414
InChI Key WZHUPCREDVWLKC-UHFFFAOYSA-N
PubChem CID 343747
IUPAC Name ethyl 2-amino-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=C(N)S1

Specifications

Melting Point 172°C to 176°C
Odor Odorless
Quantity 25 g
Synonym ethyl 2-amino-4-methylthiazole-5-carboxylate, ethyl2-amino-4-methylthiazole-5-carboxylate, 2-amino-4-methyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl 2-amino-4-methyl thiazole-5-carboxylate, ethyl 2-amino-4-methyl-thiazole-5-carboxylate, 2-amino-4-methyl-5-thiazolecarboxylic acid ethyl ester, 2-amino-4-methyl-5-ethoxycarbonyl thiazole, 2-amino-4-methyl-5-ethoxycarbonyl-thiazole, ethyl 2-amino-4-methyl-5-thiazolecarboxylate
Formula Weight 186.22
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-methylthiazole-5-carboxylate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only