missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate 98.0+%, TCI America™

Catalog No. a135510g
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A135510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A135510G Supplier TCI America Supplier No. A135510G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4506-71-2
Molecular Formula C11H15NO2S
Molecular Weight (g/mol) 225.306
MDL Number MFCD00005862
InChI Key CDYVTVLXEWMCHU-UHFFFAOYSA-N
Synonym ethyl 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid ethyl ester, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid ethyl ester, benzo b thiophene-3-carboxylic acid, 2-amino-4,5,6,7-tetrahydro-, ethyl ester, ethyl2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, methylaminotetrahydrobenzothiophenecarboxylate, 2-amino-3-carbethoxy-4,5-tetramethylenethiophene, ethyl 2-amino-4,5,6,7-tetrahydrobenzothiophene-3-carboxylate, ethyl-2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate
PubChem CID 78262
IUPAC Name ethyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC2=C1CCCC2)N

Specifications

Melting Point 115°C
Color Yellow
Quantity 10 g
Formula Weight 225.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate