missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, 96%, Thermo Scientific Chemicals

Catalog No. AAH3331903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3331903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3331903 Supplier Thermo Scientific Chemicals Supplier No. H3331903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 193140-71-5 | Molecular Formula: C5H9ClF3NO2 | Molecular Weight: 207.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 193140-71-5
Molecular Formula C5H9ClF3NO2
Molecular Weight (g/mol) 207.58
MDL Number MFCD14525479
InChI Key MNFISCBCWBUTTQ-UHFFFAOYNA-N
Synonym ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride, 2-amino-3,3,3-trifluoro-propionic acid ethyl ester hydrochloride, ethyl 3,3,3-trifluoroalaninate, ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, ethyl 3,3,3-trifluoroalaninate-hydrogen chloride 1/1
PubChem CID 45790935
SMILES Cl.CCOC(=O)C(N)C(F)(F)F

Specifications

Melting Point ∼149°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
IUPAC Name ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride
Formula Weight 207.58
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only