missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate Hydrochloride 98.0+%, TCI America™

Catalog No. E10495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10495G 5 g
E104925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10495G Supplier TCI America Supplier No. E10495G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 399017-10-8
Molecular Formula C13H14ClNO3S
Molecular Weight (g/mol) 299.769
InChI Key CTNLNGHJGMEXNQ-UHFFFAOYSA-N
Synonym 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylic Acid Ethyl Ester Hydrochloride
PubChem CID 91972097
IUPAC Name ethyl 4-methyl-2-(4-oxocyclohexa-2,5-dien-1-ylidene)-3H-1,3-thiazole-5-carboxylate;hydrochloride
SMILES CCOC(=O)C1=C(NC(=C2C=CC(=O)C=C2)S1)C.Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 299.77
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate Hydrochloride