missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2,4-Dimethylpyrrole-3-carboxylate 98.0+%, TCI America™
$73.16 - $292.62
Chemical Identifiers
| CAS | 2199-51-1 |
|---|---|
| Molecular Formula | C9H13NO2 |
| Molecular Weight (g/mol) | 167.208 |
| MDL Number | MFCD01365811 |
| InChI Key | QWSFDUPEOPMXCV-UHFFFAOYSA-N |
| Synonym | 2,4-Dimethylpyrrole-3-carboxylic Acid Ethyl Ester |
| PubChem CID | 137481 |
| IUPAC Name | ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate |
| SMILES | CCOC(=O)C1=C(NC=C1C)C |
Chemical Identifiers
| 2199-51-1 | |
| 167.208 | |
| QWSFDUPEOPMXCV-UHFFFAOYSA-N | |
| 137481 | |
| CCOC(=O)C1=C(NC=C1C)C |
| C9H13NO2 | |
| MFCD01365811 | |
| 2,4-Dimethylpyrrole-3-carboxylic Acid Ethyl Ester | |
| ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate |
Specifications
| 2199-51-1 | |
| White-Yellow | |
| C9H13NO2 | |
| 1 g | |
| QWSFDUPEOPMXCV-UHFFFAOYSA-N | |
| ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate | |
| 137481 | |
| ≥98.0% (GC) | |
| Ethyl 2,4-Dimethylpyrrole-3-carboxylate |
| 76°C | |
| 162°C | |
| MFCD01365811 | |
| 2,4-Dimethylpyrrole-3-carboxylic Acid Ethyl Ester | |
| CCOC(=O)C1=C(NC=C1C)C | |
| 167.208 | |
| 167.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No