missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2,4-Dimethylpyrrole-3-carboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 76°C |
| Color | White-Yellow |
| Boiling Point | 162°C |
| Quantity | 1 g |
| Formula Weight | 167.21 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethyl 2,4-Dimethylpyrrole-3-carboxylate |
Safety and Handling
| TSCA | No |