missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98.0+%, TCI America™

Catalog No. p-7129429
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E13111G 1 g
E13115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E13111G Supplier TCI America Supplier No. E13111G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 269409-99-6
Molecular Formula C15H21BO4
Molecular Weight (g/mol) 276.139
MDL Number MFCD03453660
InChI Key RFBZWPFBCXBBJS-UHFFFAOYSA-N
Synonym ethyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid pinacol ester, ethyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid, pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester, ethyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-carboethoxyphenylboronic acid pinacol ester, 2-ethoxycarbonyl benzeneboronic acid pinacol ester, ethyl 2-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzoate
PubChem CID 2773962
IUPAC Name ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C(=O)OCC

Specifications

Melting Point 65°C
Color White
Quantity 1 g
Formula Weight 276.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate