missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2,3,4,5-Tetrafluorobenzoyl)acetate 98.0+%, TCI America™

Catalog No. e075925g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E075925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E075925G Supplier TCI America Supplier No. E075925G
Only null left

Chemical Identifiers

CAS 94695-50-8
Molecular Formula C11H8F4O3
Molecular Weight (g/mol) 264.176
MDL Number MFCD00523020
InChI Key KWDVJYLIAJHEOW-UHFFFAOYSA-N
Synonym (2,3,4,5-Tetrafluorobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-Oxo-3-(2,3,4,5-tetrafluorophenyl)propionate, 3-Oxo-3-(2,3,4,5-tetrafluorophenyl)propionic Acid Ethyl Ester
PubChem CID 600916
IUPAC Name ethyl 3-oxo-3-(2,3,4,5-tetrafluorophenyl)propanoate
SMILES CCOC(=O)CC(=O)C1=CC(=C(C(=C1F)F)F)F

Specifications

Melting Point 48°C
Color White-Yellow
Boiling Point 100°C
Quantity 25 g
Formula Weight 264.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl (2,3,4,5-Tetrafluorobenzoyl)acetate