missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1H-pyrazole-4-carboxylate, 98%

Catalog No. AAB2535114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2535114 25 g
AAB2535106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2535114 Supplier Thermo Scientific Chemicals Supplier No. B2535114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37622-90-5 | C6H8N2O2 | 140.142 g/mol

Ethyl 1H-pyrazole-4-carboxylate is used in the preparation of isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides. Further, it acts as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1H-pyrazole-4-carboxylate is used in the preparation of isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides. Further, it acts as an intermediate in organic synthesis.

Solubility
Miscible with acetone.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 37622-90-5
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00010844
InChI Key KACZQOKEFKFNDB-UHFFFAOYSA-N
PubChem CID 142179
IUPAC Name ethyl 1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=CNN=C1

Specifications

Melting Point 75°C to 80°C
Boiling Point 138°C to 140°C (3 mmHg)
Quantity 25 g
Synonym ethyl 4-pyrazolecarboxylate, ethyl pyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid ethyl ester, 4-ethoxycarbonylpyrazole, 1h-pyrazole-4-carboxylic acid, ethyl ester, 4-pyrazolecarboxylic acid ethyl ester, ethyl4-pyrazolecarboxylate, zlchem 132, pubchem21939, acmc-209iug
Solubility Information Miscible with acetone.
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material Ethyl 1H-pyrazole-4-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only