missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (11bR)-4-Amino-2,6-bis(3,5-di-tert-butylphenyl)-4,5-dihydro-3H-cyclohepta[1,2-a:7,6-a']dinaphthalene-4-carbo 97+%, TCI America™
Supplier: TCI America E126750MG
Specifications
| Ethyl (11bR)-4-Amino-2,6-bis(3,5-di-tert-butylphenyl)-4,5-dihydro-3H-cyclohepta[1,2-a:7,6-a′]d | |
| White | |
| 50 mg | |
| 758.10 | |
| Crystalline Powder |
| 1678540-23-2 | |
| C54H63NO2 | |
| (11bR)-4-Amino-2,6-bis(3,5-di-tert-butylphenyl)-4,5-dihydro-3H-cyclohepta[1,2-a:7,6-a′C]dinaphthalene-4-carboxylic Acid Ethyl Ester | |
| ≥97.0% (HPLC) |
Chemical Identifiers
| 1678540-23-2 | |
| C54H63NO2 | |
| (11bR)-4-Amino-2,6-bis(3,5-di-tert-butylphenyl)-4,5-dihydro-3H-cyclohepta[1,2-a:7,6-a′C]dinaphthalene-4-carboxylic Acid Ethyl Ester |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator