missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 10-Undecenoate 97.0+%, TCI America™
Supplier: TCI America E0771250G
Specifications
| Ethyl 10-Undecenoate | |
| -38°C | |
| 259°C | |
| MFCD00009220 | |
| 10-Undecenoic Acid Ethyl Ester | |
| CCOC(=O)CCCCCCCCC=C | |
| 212.333 | |
| 212.33 | |
| Liquid |
| 692-86-4 | |
| Yellow | |
| C13H24O2 | |
| 250 g | |
| FXNFFCMITPHEIT-UHFFFAOYSA-N | |
| ethyl undec-10-enoate | |
| 12729 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 692-86-4 | |
| 212.333 | |
| FXNFFCMITPHEIT-UHFFFAOYSA-N | |
| 12729 | |
| CCOC(=O)CCCCCCCCC=C |
| C13H24O2 | |
| MFCD00009220 | |
| 10-Undecenoic Acid Ethyl Ester | |
| ethyl undec-10-enoate |
Safety and Handling
EINECSNumber : (2)-0798
RTECSNumber : YQ2978400
TSCA : Yes