missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 10-Undecenoate 97.0+%, TCI America™

Catalog No. E077125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E077125G 25 g
E0771250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E077125G Supplier TCI America Supplier No. E077125G
Only null left

Chemical Identifiers

CAS 692-86-4
Molecular Formula C13H24O2
Molecular Weight (g/mol) 212.333
MDL Number MFCD00009220
InChI Key FXNFFCMITPHEIT-UHFFFAOYSA-N
Synonym 10-Undecenoic Acid Ethyl Ester
PubChem CID 12729
IUPAC Name ethyl undec-10-enoate
SMILES CCOC(=O)CCCCCCCCC=C

Specifications

Melting Point -38°C
Color Yellow
Boiling Point 259°C
Quantity 25 g
Formula Weight 212.33
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 10-Undecenoate
EINECSNumber (2)-0798
RTECSNumber YQ2978400
TSCA Yes