missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH3284703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3284703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3284703 Supplier Thermo Scientific Chemicals Supplier No. H3284703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1040683-08-6 | Molecular Formula: C8H11F3O2 | Molecular Weight: 196.169 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1040683-08-6
Molecular Formula C8H11F3O2
Molecular Weight (g/mol) 196.169
MDL Number MFCD10699116
InChI Key AYUISFJYNKIHDD-UHFFFAOYSA-N
PubChem CID 28309116
IUPAC Name ethyl 1-(trifluoromethyl)cyclobutane-1-carboxylate
SMILES CCOC(=O)C1(CCC1)C(F)(F)F

Specifications

Boiling Point 61°C to 63°C (20 mmHg)
Quantity 1 g
Synonym ethyl 1-trifluoromethyl cyclobutanecarboxylate, 1-trifluoromethyl cyclobutanecarboxylic acid ethyl ester, ethyl 1-trifluoromethyl cyclobutane-1-carboxylate, ethyl1-trifluoromethyl cyclobutanecarboxylate, 1-trifluoromethyl cyclobutanecarboxylic acidethyl ester
Formula Weight 196.17
Percent Purity 97%
Chemical Name or Material Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only