missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate 98.0+%, TCI America™

Catalog No. E09545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09545G 5 g
E095425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09545G Supplier TCI America Supplier No. E09545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130250-54-3
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD04116274
InChI Key YCXCRFGBFZTUSU-UHFFFAOYSA-N
Synonym ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester
PubChem CID 357727
IUPAC Name 1-O-tert-butyl 3-O-ethyl piperidine-1,3-dicarboxylate
SMILES CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C

Specifications

Melting Point 37°C
Color Yellow
Boiling Point 93°C
Quantity 5 g
Formula Weight 257.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate