missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-Propenyl Ether (cis- and trans- mixture) 98.0+%, TCI America™
Supplier: TCI America E1052100ML
Specifications
| Ethyl 1-Propenyl Ether (cis- and trans- mixture) | |
| Colorless | |
| C5H10O | |
| 100 mL | |
| 1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether | |
| CCO\C=C/C | |
| 86.13 | |
| 86.13 | |
| Liquid |
| 928-55-2 | |
| 70°C | |
| MFCD00015184 | |
| 3271 | |
| XDHOEHJVXXTEDV-HYXAFXHYSA-N | |
| (1Z)-1-ethoxyprop-1-ene | |
| 5365091 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 928-55-2 | |
| 86.13 | |
| XDHOEHJVXXTEDV-HYXAFXHYSA-N | |
| 5365091 | |
| CCO\C=C/C |
| C5H10O | |
| MFCD00015184 | |
| 1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether | |
| (1Z)-1-ethoxyprop-1-ene |
Safety and Handling
EINECSNumber : (2)-4052
RTECSNumber : KO0525000
TSCA : No