missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Propenyl Ether (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. E105225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E105225ML 25 mL
E1052100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E105225ML Supplier TCI America Supplier No. E105225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 928-55-2
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00015184
InChI Key XDHOEHJVXXTEDV-HYXAFXHYSA-N
Synonym 1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
PubChem CID 5365091
IUPAC Name (1Z)-1-ethoxyprop-1-ene
SMILES CCO\C=C/C

Specifications

Color Colorless
Boiling Point 70°C
Quantity 25 mL
UN Number 3271
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1-Propenyl Ether (cis- and trans- mixture)
EINECSNumber (2)-4052
RTECSNumber KO0525000
TSCA No