missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Piperazinecarboxylate 96.0+%, TCI America™

Catalog No. P134925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P134925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P134925G Supplier TCI America Supplier No. P134925G

Specifications

CAS 120-43-4
Color Yellow
Boiling Point 237°C
Molecular Formula C7H14N2O2
Quantity 25 g
MDL Number MFCD00005964
Synonym ethyl n-piperazinecarboxylate, ethyl 1-piperazinecarboxylate, n-carbethoxypiperazine, 1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-piperazinecarboxylic acid, ethyl ester, n-ethoxycarbonyl piperazine, ethylcarbonyl piperazine, 1-ethoxycarbonyl piperazine, ethyl 1-piperazinocarboxylate
InChI Key LNOQURRKNJKKBU-UHFFFAOYSA-N
SMILES CCOC(=O)N1CCNCC1
IUPAC Name ethyl piperazine-1-carboxylate
Molecular Weight (g/mol) 158.201
PubChem CID 8431
Formula Weight 158.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1-Piperazinecarboxylate
Show More Show Less
RTECSNumber TL1378000
TSCA Yes