missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-methyl-5-thien-2-yl-1H-pyrazole-3-carboxylate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC47123DA

Catalog No. CC47123DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl 1-methyl-5-thien-2-yl-1H-pyrazole-3-carboxylate
96.5
97%
MFCD08060536
ethyl 1-methyl-5-thien-2-yl-1h-pyrazole-3-carboxylate, ethyl 1-methyl-5-thiophen-2-yl pyrazole-3-carboxylate, ethyl 1-methyl-5-2-thienyl pyrazole-3-carboxylate, ethyl 1-methyl-5-thiophen-2-yl-1h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylicacid,1-methyl-5-2-thienyl-,ethyl ester, 1h-pyrazole-3-carboxylicacid, 1-methyl-5-2-thienyl-, ethyl ester
CCOC(=O)C1=NN(C)C(=C1)C1=CC=CS1
236.29
236.29
868755-60-6
100.0
C11H12N2O2S
1g
KIYMTNJEKXVPSQ-UHFFFAOYSA-N
ethyl 1-methyl-5-thiophen-2-ylpyrazole-3-carboxylate
7537650
97%

Chemical Identifiers

868755-60-6
236.29
KIYMTNJEKXVPSQ-UHFFFAOYSA-N
7537650
CCOC(=O)C1=NN(C)C(=C1)C1=CC=CS1
C11H12N2O2S
MFCD08060536
ethyl 1-methyl-5-thien-2-yl-1h-pyrazole-3-carboxylate, ethyl 1-methyl-5-thiophen-2-yl pyrazole-3-carboxylate, ethyl 1-methyl-5-2-thienyl pyrazole-3-carboxylate, ethyl 1-methyl-5-thiophen-2-yl-1h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylicacid,1-methyl-5-2-thienyl-,ethyl ester, 1h-pyrazole-3-carboxylicacid, 1-methyl-5-2-thienyl-, ethyl ester
ethyl 1-methyl-5-thiophen-2-ylpyrazole-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions