missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Methyl-3-piperidinecarboxylate 95.0+%, TCI America™

Catalog No. E08635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08635G 5 g
E086325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08635G Supplier TCI America Supplier No. E08635G

Chemical Identifiers

CAS 5166-67-6
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00006496
InChI Key VFJJNMLPRDRTCO-UHFFFAOYSA-N
Synonym ethyl 1-methylnipecotate, ethyl n-methylpiperidine-3-carboxylate, ethyl 1-methyl-3-piperidinecarboxylate, 1-methyl-piperidine-3-carboxylic acid ethyl ester, 1-methylpiperidine-3-carboxylic acid ethyl ester, 1-methyl-3-piperidinecarboxylic acid ethyl ester, 3-piperidinecarboxylic acid, 1-methyl-, ethyl ester, ethyl-1-methylpiperidin-3-carboxylat, pubchem11774, acmc-209kvb
PubChem CID 97981
IUPAC Name ethyl 1-methylpiperidine-3-carboxylate
SMILES CCOC(=O)C1CCCN(C1)C

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 171.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl 1-Methyl-3-piperidinecarboxylate