missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals

Catalog No. aah6280403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6280403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6280403 Supplier Thermo Scientific Chemicals Supplier No. H6280403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1075-82-7 | Molecular Formula: C9H16O3 | Molecular Weight: 172.224 g/mol

Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate is employed as a intermediate for chemical research and synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate is employed as a intermediate for chemical research and synthetic building block.

Notes
Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 1075-82-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD12022629
InChI Key AUUICUZWYQYTNO-UHFFFAOYSA-N
Synonym ethyl 1-hydroxymethyl cyclopentanecarboxylate, ethyl 1-hydroxymethyl cyclopentane-1-carboxylate, 1-hydroxymethyl-cyclopentanecarboxylic acid ethyl ester
PubChem CID 46172884
IUPAC Name ethyl 1-(hydroxymethyl)cyclopentane-1-carboxylate
SMILES CCOC(=O)C1(CCCC1)CO

Specifications

Quantity 1 g
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl 1-(hydroxymethyl)cyclopentanecarboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only