missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-(hydroxymethyl)cyclobutanecarboxylate, 97%
CAS: 1195-81-9 | C8H14O3 | 158.197 g/mol
Supplier: Thermo Scientific Chemicals H6256603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl 1-(hydroxymethyl)cyclobutanecarboxylate | |
| C8H14O3 | |
| 1 g | |
| PWMQFMMZBJUHID-UHFFFAOYSA-N | |
| ethyl 1-(hydroxymethyl)cyclobutane-1-carboxylate | |
| 15670506 | |
| 97% |
| 1195-81-9 | |
| MFCD11707219 | |
| ethyl 1-hydroxymethyl cyclobutane-1-carboxylate, ethyl 1-hydroxymethylcyclobutanecarboxylate, ethyl 1-hydroxymethyl cyclobutanecarboxylate, cyclobutanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl-cyclobutanecarboxylate, 1-hydroxymethyl-cyclobutanecarboxylic acid ethyl ester | |
| CCOC(=O)C1(CCC1)CO | |
| 158.197 | |
| 158.19 |
Chemical Identifiers
| 1195-81-9 | |
| 158.197 | |
| PWMQFMMZBJUHID-UHFFFAOYSA-N | |
| 15670506 | |
| CCOC(=O)C1(CCC1)CO |
| C8H14O3 | |
| MFCD11707219 | |
| ethyl 1-hydroxymethyl cyclobutane-1-carboxylate, ethyl 1-hydroxymethylcyclobutanecarboxylate, ethyl 1-hydroxymethyl cyclobutanecarboxylate, cyclobutanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl-cyclobutanecarboxylate, 1-hydroxymethyl-cyclobutanecarboxylic acid ethyl ester | |
| ethyl 1-(hydroxymethyl)cyclobutane-1-carboxylate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only