missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate 98.0+%, TCI America™

Catalog No. E08885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08885G 5 g
E088825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08885G Supplier TCI America Supplier No. E08885G
Only null left

Chemical Identifiers

CAS 100501-62-0
Molecular Formula C14H12F3NO3
Molecular Weight (g/mol) 299.25
MDL Number MFCD00276022
InChI Key LWLLHOVWIFISMG-UHFFFAOYSA-N
Synonym ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid ethyl ester, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxohydroquinoline-3-carboxylic acid ethyl ester, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate dry wt. , may cont. up to water, acmc-1brcg, ksc499a1t, ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylate, neopentyl glycol-tetrahydrofuran copolymer, 2,2-dimethyl-1,3-propanediol-tetramethylene oxide copolymer
PubChem CID 5140913
IUPAC Name ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN(CC)C2=C(F)C(F)=C(F)C=C2C1=O

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 5 g
Formula Weight 299.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-Ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate