missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate 98.0+%, TCI America™
$52.19 - $171.49
Chemical Identifiers
| CAS | 112811-71-9 |
|---|---|
| Molecular Formula | C16H15F2NO4 |
| Molecular Weight (g/mol) | 323.296 |
| MDL Number | MFCD05864419 |
| InChI Key | XPAOPAPDCRLMTR-UHFFFAOYSA-N |
| Synonym | 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester |
| PubChem CID | 10758239 |
| IUPAC Name | ethyl 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxoquinoline-3-carboxylate |
| SMILES | CCOC(=O)C1=CN(C2=C(C(=C(C=C2C1=O)F)F)OC)C3CC3 |
Chemical Identifiers
| 112811-71-9 | |
| 323.296 | |
| XPAOPAPDCRLMTR-UHFFFAOYSA-N | |
| 10758239 | |
| CCOC(=O)C1=CN(C2=C(C(=C(C=C2C1=O)F)F)OC)C3CC3 |
| C16H15F2NO4 | |
| MFCD05864419 | |
| 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester | |
| ethyl 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxoquinoline-3-carboxylate |
Specifications
| 112811-71-9 | |
| White | |
| MFCD05864419 | |
| 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester | |
| CCOC(=O)C1=CN(C2=C(C(=C(C=C2C1=O)F)F)OC)C3CC3 | |
| 323.296 | |
| 323.30 | |
| Crystalline Powder |
| 185°C | |
| C16H15F2NO4 | |
| 1 g | |
| XPAOPAPDCRLMTR-UHFFFAOYSA-N | |
| ethyl 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxoquinoline-3-carboxylate | |
| 10758239 | |
| ≥98.0% (GC) | |
| Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate |
Safety and Handling
EINECSNumber : (5)-6708
TSCA : No
Recommended Storage : Freezer