missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate 98.0+%, TCI America™

Catalog No. E11475G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11475G 5 g
E11471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. E11475G missing translation for 'mfr' TCI America missing translation for 'supplierNo' E11475G

Chemical Identifiers

CAS 112811-71-9
Molecular Formula C16H15F2NO4
Molecular Weight (g/mol) 323.296
MDL Number MFCD05864419
InChI Key XPAOPAPDCRLMTR-UHFFFAOYSA-N
Synonym 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester
PubChem CID 10758239
IUPAC Name ethyl 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxoquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN(C2=C(C(=C(C=C2C1=O)F)F)OC)C3CC3

Specifications

Melting Point 185°C
Color White
Quantity 5 g
Formula Weight 323.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate
EINECSNumber (5)-6708
TSCA No
Recommended Storage Freezer