missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Boc-DL-nipecotate, 97%, Thermo Scientific Chemicals

Catalog No. p-7031578
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3270406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3270406 Supplier Thermo Scientific Chemicals Supplier No. H3270406

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130250-54-3
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD04116274
InChI Key YCXCRFGBFZTUSU-UHFFFAOYSA-N
Synonym ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester
PubChem CID 357727
IUPAC Name 1-O-tert-butyl 3-O-ethyl piperidine-1,3-dicarboxylate
SMILES CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C

Specifications

Melting Point 31°C to 35°C
Boiling Point 95°C (0.5 mmHg)
Quantity 5 g
Sensitivity Light and moisture sensitive
Formula Weight 257.33
Percent Purity 97%
Chemical Name or Material Ethyl 1-Boc-DL-nipecotate
Hazard Category H302-H317-H319
Hazard Statement GHS H Statement
H301-H319-H317
Toxic if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only