missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Benzylpyrrole-3-carboxylate 95.0+%, TCI America™

Catalog No. B22841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22841G 1 g
B22845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22841G Supplier TCI America Supplier No. B22841G
Only null left

Chemical Identifiers

CAS 128259-47-2
Molecular Formula C14H15NO2
Molecular Weight (g/mol) 229.279
MDL Number MFCD06797056
InChI Key KSZAVNAKDOXZGJ-UHFFFAOYSA-N
Synonym 1-Benzylpyrrole-3-carboxylic Acid Ethyl Ester
PubChem CID 14667729
IUPAC Name ethyl 1-benzylpyrrole-3-carboxylate
SMILES CCOC(=O)C1=CN(C=C1)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 229.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1-Benzylpyrrole-3-carboxylate
TSCA No
Recommended Storage Refrigerator