missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Aminocyclopropanecarboxylate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7129233
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09371G 1 g
E09375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09371G Supplier TCI America Supplier No. E09371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42303-42-4
Molecular Formula C6H12ClNO2
Molecular Weight (g/mol) 165.617
MDL Number MFCD00190747
InChI Key XFNUTZWASODOQK-UHFFFAOYSA-N
Synonym ethyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet.hcl, ethyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1-amino-cyclopropyl-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet hcl, 1-aminocyclopropane-1-carboxylic acid ethyl ester, acpc-oet-hcl, h-acpc-oet?cl, pubchem14733
PubChem CID 386203
IUPAC Name ethyl 1-aminocyclopropane-1-carboxylate;hydrochloride
SMILES CCOC(=O)C1(CC1)N.Cl

Specifications

Melting Point 110°C
Color White
Quantity 1 g
Formula Weight 165.62
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-Aminocyclopropanecarboxylate Hydrochloride