missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-aminocyclopropanecarboxylate hydrochloride, 98%

Catalog No. AC432510250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432510250 25 g Glass bottle
AC432510050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432510250 Supplier Thermo Scientific Chemicals Supplier No. 432510250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42303-42-4 | C6H11NO2·ClH | 165.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42303-42-4
Molecular Formula C6H11NO2·ClH
Molecular Weight (g/mol) 165.62
MDL Number MFCD00190747
InChI Key XFNUTZWASODOQK-UHFFFAOYSA-N
Synonym ethyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet.hcl, ethyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1-amino-cyclopropyl-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet hcl, 1-aminocyclopropane-1-carboxylic acid ethyl ester, acpc-oet-hcl, h-acpc-oet?cl, pubchem14733
PubChem CID 386203
IUPAC Name ethyl 1-aminocyclopropane-1-carboxylate;hydrochloride
SMILES CCOC(=O)C1(CC1)N.Cl

Specifications

Melting Point 113.0°C to 114.0°C
Color White
Elemental Analysis 42.41 to 45.48% C, 8.24 to 8.66% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Formula Weight 165.62
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-aminocyclopropanecarboxylate hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only