missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,3-dithiolane-2-carboxylate, 99%, Thermo Scientific™

$134.18 - $134.18

Chemical Identifiers

CAS 20461-99-8
Molecular Formula C6H10O2S2
Molecular Weight (g/mol) 178.26
MDL Number MFCD00005411
InChI Key OMCSHTHLIQOHDD-UHFFFAOYSA-N
Synonym 2-carboethoxydithiolane, ethyl1,3-dithiolane-2-carboxylate, 1,3-dithiolane-2-carboxylic acid ethyl ester, acmc-209faj, 2-carboethoxy-1,3-dithiolane, omcshthliqohdd-uhfffaoysa, 1,3-dithiolane-2-carboxylicacid,ethyl ester, 1,3-dithiolane-2-carboxylicacid, ethyl ester, 1,3-dithiolane-2-carboxylic acid, ethyl ester, ethyl 1,3-dithiolane-2-carboxylate
PubChem CID 88551
IUPAC Name ethyl 1,3-dithiolane-2-carboxylate
SMILES CCOC(=O)C1SCCS1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC209990050
SDS View Documents
Thermo Scientific Chemicals
209990050
5 mL Glass bottle
Each for $134.18
Only null left
 
Description

Description

Specifications

Chemical Identifiers

20461-99-8
178.26
OMCSHTHLIQOHDD-UHFFFAOYSA-N
88551
CCOC(=O)C1SCCS1
C6H10O2S2
MFCD00005411
2-carboethoxydithiolane, ethyl1,3-dithiolane-2-carboxylate, 1,3-dithiolane-2-carboxylic acid ethyl ester, acmc-209faj, 2-carboethoxy-1,3-dithiolane, omcshthliqohdd-uhfffaoysa, 1,3-dithiolane-2-carboxylicacid,ethyl ester, 1,3-dithiolane-2-carboxylicacid, ethyl ester, 1,3-dithiolane-2-carboxylic acid, ethyl ester, ethyl 1,3-dithiolane-2-carboxylate
ethyl 1,3-dithiolane-2-carboxylate

Specifications

20461-99-8
1.2400g/mL
110°C
98.5% min. (GC)
C6H10O2S2
MFCD00005411
19, II, 290
2-carboethoxydithiolane, ethyl1,3-dithiolane-2-carboxylate, 1,3-dithiolane-2-carboxylic acid ethyl ester, acmc-209faj, 2-carboethoxy-1,3-dithiolane, omcshthliqohdd-uhfffaoysa, 1,3-dithiolane-2-carboxylicacid,ethyl ester, 1,3-dithiolane-2-carboxylicacid, ethyl ester, 1,3-dithiolane-2-carboxylic acid, ethyl ester, ethyl 1,3-dithiolane-2-carboxylate
CCOC(=O)C1SCCS1
178.26
178.26
Liquid
Yellow
85°C (0.1 mmHg)
Authentic
Glass bottle
1.5380 to 1.54
5 mL
1.24
OMCSHTHLIQOHDD-UHFFFAOYSA-N
ethyl 1,3-dithiolane-2-carboxylate
88551
99%
Ethyl 1, 3-dithiolane-2-carboxylate, 99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 243-837-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only