missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,1,2,3,3,3-Hexafluoropropyl Ether 98.0+%, TCI America™

Catalog No. E10205G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10205G Supplier TCI America Supplier No. E10205G
Only null left

Specifications

CAS 380-34-7
Color Yellow
Boiling Point 65°C
Molecular Formula C5H6F6O
Quantity 5 g
MDL Number MFCD00236122
UN Number 3271
InChI Key DMECHFLLAQSVAD-UHFFFAOYSA-N
SMILES CCOC(C(C(F)(F)F)F)(F)F
IUPAC Name 1-ethoxy-1,1,2,3,3,3-hexafluoropropane
Molecular Weight (g/mol) 196.092
PubChem CID 2775036
Formula Weight 196.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1,1,2,3,3,3-Hexafluoropropyl Ether
Show More Show Less