missing translation for 'onlineSavingsMsg'
Learn More

Ethoxytrimethylsilane, 95%

Catalog No. p-3757810
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313820250 25 g Glass bottle
AC313821000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC313820250 Supplier Thermo Scientific Chemicals Supplier No. 313820250
Only null left

CAS: 1825-62-3 | C5H14OSi | 118.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1825-62-3
Molecular Formula C5H14OSi
Molecular Weight (g/mol) 118.25
MDL Number MFCD00009069
InChI Key RSIHJDGMBDPTIM-UHFFFAOYSA-N
Synonym trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
PubChem CID 15772
IUPAC Name ethoxy(trimethyl)silane
SMILES CCO[Si](C)(C)C

Specifications

Color Colorless
Density 0.7500g/mL
Boiling Point 75°C to 76°C
Flash Point -18°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula Si(CH3)3OC2H5
Refractive Index 1.3730 to 1.375
Quantity 25 g
Specific Gravity 0.75
Solubility Information Solubility in water: reacts with water
Formula Weight 118.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethoxytrimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-370-6

RUO – Research Use Only