missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethoxyquin, ≥90%, MP Biomedicals™
$188.72 - $921.57
Chemical Identifiers
| CAS | 91-53-2 |
|---|---|
| Molecular Formula | C14H19NO |
| Molecular Weight (g/mol) | 217.312 |
| MDL Number | MFCD00023883 |
| InChI Key | DECIPOUIJURFOJ-UHFFFAOYSA-N |
| Synonym | ethoxyquin, 6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, ethoxyquine, santoquin, santoquine, niflex, antioxidant ec, santoflex a, santoflex aw, stop-scald |
| PubChem CID | 3293 |
| ChEBI | CHEBI:77323 |
| IUPAC Name | 6-ethoxy-2,2,4-trimethyl-1H-quinoline |
| SMILES | CCOC1=CC2=C(C=C1)NC(C=C2C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
ICN15796380
|
MP Biomedicals Inc
0215796380 |
100 g |
Each for $188.72
|
|
|||||
|
ICN15796383
|
MP Biomedicals Inc
0215796383 |
250 g |
Each for $350.33
|
|
|||||
|
ICN15796391
|
MP Biomedicals Inc
0215796391 |
1 kg |
Each for $921.57
|
|
|||||
Chemical Identifiers
| 91-53-2 | |
| 217.312 | |
| DECIPOUIJURFOJ-UHFFFAOYSA-N | |
| 3293 | |
| 6-ethoxy-2,2,4-trimethyl-1H-quinoline |
| C14H19NO | |
| MFCD00023883 | |
| ethoxyquin, 6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, ethoxyquine, santoquin, santoquine, niflex, antioxidant ec, santoflex a, santoflex aw, stop-scald | |
| CHEBI:77323 | |
| CCOC1=CC2=C(C=C1)NC(C=C2C)(C)C |
Specifications
| 91-53-2 | |
| 1.030g/mL | |
| C14H19NO | |
| 100 g | |
| DECIPOUIJURFOJ-UHFFFAOYSA-N | |
| 6-ethoxy-2,2,4-trimethyl-1H-quinoline | |
| 3293 | |
| 217.3 | |
| Ethoxyquin |
| Brown | |
| ≥90% | |
| MFCD00023883 | |
| ethoxyquin, 6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, ethoxyquine, santoquin, santoquine, niflex, antioxidant ec, santoflex a, santoflex aw, stop-scald | |
| CCOC1=CC2=C(C=C1)NC(C=C2C)(C)C | |
| 217.312 | |
| CHEBI:77323 | |
| Viscous Liquid |
Safety and Handling
EINECSNumber : 202-075-7
Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated. Store under nitrogen.