missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyquin, ≥90%, MP Biomedicals™

Catalog No. ICN15796380
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15796380 100 g
ICN15796383 250 g
ICN15796391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15796380 Supplier MP Biomedicals Inc Supplier No. 0215796380
Only null left

Chemical Identifiers

CAS 91-53-2
Molecular Formula C14H19NO
Molecular Weight (g/mol) 217.312
MDL Number MFCD00023883
InChI Key DECIPOUIJURFOJ-UHFFFAOYSA-N
Synonym ethoxyquin, 6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, ethoxyquine, santoquin, santoquine, niflex, antioxidant ec, santoflex a, santoflex aw, stop-scald
PubChem CID 3293
ChEBI CHEBI:77323
IUPAC Name 6-ethoxy-2,2,4-trimethyl-1H-quinoline
SMILES CCOC1=CC2=C(C=C1)NC(C=C2C)(C)C

Specifications

Color Brown
Density 1.030g/mL
Assay Percent Range ≥90%
Quantity 100 g
Formula Weight 217.3
Physical Form Viscous Liquid
Chemical Name or Material Ethoxyquin
EINECSNumber 202-075-7
Recommended Storage Store at Room Temperature (15 to 30°C). Store Desiccated. Store under nitrogen.