Learn More
Ethoxymethylenemalononitrile, 97+%
CAS: 123-06-8 | C6H6N2O | 122.127 g/mol
$98.49 - $215.37
Chemical Identifiers
| CAS | 123-06-8 |
|---|---|
| Molecular Formula | C6H6N2O |
| Molecular Weight (g/mol) | 122.127 |
| MDL Number | MFCD00001854 |
| InChI Key | OEICGMPRFOJHKO-UHFFFAOYSA-N |
| Synonym | ethoxymethylenemalononitrile, ethoxymethylene malononitrile, propanedinitrile, ethoxymethylene, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, 2-cyano-3-ethoxyacrylonitrile, ethoxymethylene propanedinitrile, usaf a-9230, usaf kf-10 |
| PubChem CID | 67152 |
| IUPAC Name | 2-(ethoxymethylidene)propanedinitrile |
| SMILES | CCOC=C(C#N)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1441214
|
Thermo Scientific Chemicals
A1441214 |
25 g |
Each for $98.49
|
|
|||||
|
AAA1441222
|
Thermo Scientific Chemicals
A1441222 |
100 g |
Each for $215.37
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 123-06-8 | |
| 122.127 | |
| OEICGMPRFOJHKO-UHFFFAOYSA-N | |
| 67152 | |
| CCOC=C(C#N)C#N |
| C6H6N2O | |
| MFCD00001854 | |
| ethoxymethylenemalononitrile, ethoxymethylene malononitrile, propanedinitrile, ethoxymethylene, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, 2-cyano-3-ethoxyacrylonitrile, ethoxymethylene propanedinitrile, usaf a-9230, usaf kf-10 | |
| 2-(ethoxymethylidene)propanedinitrile |
Specifications
| 123-06-8 | |
| 160°C (12 mmHg) | |
| C6H6N2O | |
| 25 g | |
| 1634241 | |
| OEICGMPRFOJHKO-UHFFFAOYSA-N | |
| 2-(ethoxymethylidene)propanedinitrile | |
| 67152 | |
| ≥97% |
| 62°C to 68°C | |
| 155°C (311°F) | |
| MFCD00001854 | |
| UN3439 | |
| ethoxymethylenemalononitrile, ethoxymethylene malononitrile, propanedinitrile, ethoxymethylene, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, 2-cyano-3-ethoxyacrylonitrile, ethoxymethylene propanedinitrile, usaf a-9230, usaf kf-10 | |
| CCOC=C(C#N)C#N | |
| 122.127 | |
| 122.13 | |
| Ethoxymethylenemalononitrile |
Safety and Handling
GHS H Statement
H301-H311-H334-H317
Toxic if swallowed.
Toxic in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 204-597-0
RTECSNumber : OO3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only