missing translation for 'onlineSavingsMsg'
Learn More

Ethoxymethylenemalononitrile, 97+%

Catalog No. AAA1441214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1441214 25 g
AAA1441222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1441214 Supplier Thermo Scientific Chemicals Supplier No. A1441214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 123-06-8 | C6H6N2O | 122.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-06-8
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00001854
InChI Key OEICGMPRFOJHKO-UHFFFAOYSA-N
Synonym ethoxymethylenemalononitrile, ethoxymethylene malononitrile, propanedinitrile, ethoxymethylene, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, 2-cyano-3-ethoxyacrylonitrile, ethoxymethylene propanedinitrile, usaf a-9230, usaf kf-10
PubChem CID 67152
IUPAC Name 2-(ethoxymethylidene)propanedinitrile
SMILES CCOC=C(C#N)C#N

Specifications

Melting Point 62°C to 68°C
Boiling Point 160°C (12 mmHg)
Flash Point 155°C (311°F)
Quantity 25 g
UN Number UN3439
Beilstein 1634241
Formula Weight 122.13
Percent Purity ≥97%
Chemical Name or Material Ethoxymethylenemalononitrile
Hazard Category H301+H311+H331-H317-H334-H335
Hazard Statement GHS H Statement
H301-H311-H334-H317
Toxic if swallowed.
Toxic in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P311-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 204-597-0
RTECSNumber OO3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only