missing translation for 'onlineSavingsMsg'
Learn More

(Ethoxymethylene)malononitrile 98.0+%, TCI America™

Catalog No. E064925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E064925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E064925G Supplier TCI America Supplier No. E064925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-06-8
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00001854
InChI Key OEICGMPRFOJHKO-UHFFFAOYSA-N
Synonym ethoxymethylenemalononitrile, ethoxymethylene malononitrile, propanedinitrile, ethoxymethylene, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, 2-cyano-3-ethoxyacrylonitrile, ethoxymethylene propanedinitrile, usaf a-9230, usaf kf-10
PubChem CID 67152
IUPAC Name 2-(ethoxymethylidene)propanedinitrile
SMILES CCOC=C(C#N)C#N

Specifications

Melting Point 66°C
Color Yellow
Boiling Point 157°C
Quantity 25 g
UN Number 3439
Formula Weight 122.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (Ethoxymethylene)malononitrile
EINECSNumber (9)-0247
RTECSNumber OO3850000
TSCA Yes