missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethoxycarbonyl isocyanate, 90%, tech.
CAS: 19617-43-7 | C4H5NO3 | 115.09 g/mol
$213.55 - $700.58
Chemical Identifiers
| CAS | 19617-43-7 |
|---|---|
| Molecular Formula | C4H5NO3 |
| Molecular Weight (g/mol) | 115.09 |
| InChI Key | VMVZGGPZNHFGKS-UHFFFAOYSA-N |
| Synonym | ethoxycarbonyl isocyanate, ethyl isocyanatoformate, ethyl carbonisocyanatidate, ethyl n-carbonylcarbamate, ethoxycarbonylisocyanate, carbonisocyanatidic acid, ethyl ester, ethyl isocyanatooate, ethylisocyanatoformate, acmc-20al2o, carbonisocyanatidicacid,ethylester |
| PubChem CID | 300166 |
| IUPAC Name | ethyl N-(oxomethylidene)carbamate |
| SMILES | CCOC(=O)N=C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC314310010
|
Thermo Scientific Chemicals
314310010 |
1 g | Glass bottle |
Each for $213.55
|
|
||||
|
AC314310050
|
Thermo Scientific Chemicals
314310050 |
5 g | Glass bottle |
Each for $700.58
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19617-43-7 | |
| 115.09 | |
| ethoxycarbonyl isocyanate, ethyl isocyanatoformate, ethyl carbonisocyanatidate, ethyl n-carbonylcarbamate, ethoxycarbonylisocyanate, carbonisocyanatidic acid, ethyl ester, ethyl isocyanatooate, ethylisocyanatoformate, acmc-20al2o, carbonisocyanatidicacid,ethylester | |
| ethyl N-(oxomethylidene)carbamate |
| C4H5NO3 | |
| VMVZGGPZNHFGKS-UHFFFAOYSA-N | |
| 300166 | |
| CCOC(=O)N=C=O |
Specifications
| 19617-43-7 | |
| 1.1100g/mL | |
| 35°C | |
| 88% min. (GC) | |
| C4H5NO3 | |
| 1.4060 to 1.4090 | |
| 1.11 | |
| VMVZGGPZNHFGKS-UHFFFAOYSA-N | |
| ethyl N-(oxomethylidene)carbamate | |
| 300166 | |
| 90% | |
| Ethoxycarbonyl isocyanate |
| Colorless | |
| 25.0°C (10.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| C2H5O2CNCO | |
| 1 g | |
| ethoxycarbonyl isocyanate, ethyl isocyanatoformate, ethyl carbonisocyanatidate, ethyl n-carbonylcarbamate, ethoxycarbonylisocyanate, carbonisocyanatidic acid, ethyl ester, ethyl isocyanatooate, ethylisocyanatoformate, acmc-20al2o, carbonisocyanatidicacid,ethylester | |
| CCOC(=O)N=C=O | |
| 115.09 | |
| 115.09 | |
| Liquid |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only