missing translation for 'onlineSavingsMsg'
Learn More

Ethoxycarbonyl isocyanate, 90%, tech.

Catalog No. AC314310010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314310010 1 g Glass bottle
AC314310050 5 g Glass bottle
AC314310250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC314310010 Supplier Thermo Scientific Chemicals Supplier No. 314310010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19617-43-7 | C4H5NO3 | 115.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19617-43-7
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.09
InChI Key VMVZGGPZNHFGKS-UHFFFAOYSA-N
Synonym ethoxycarbonyl isocyanate, ethyl isocyanatoformate, ethyl carbonisocyanatidate, ethyl n-carbonylcarbamate, ethoxycarbonylisocyanate, carbonisocyanatidic acid, ethyl ester, ethyl isocyanatooate, ethylisocyanatoformate, acmc-20al2o, carbonisocyanatidicacid,ethylester
PubChem CID 300166
IUPAC Name ethyl N-(oxomethylidene)carbamate
SMILES CCOC(=O)N=C=O

Specifications

Color Colorless
Density 1.1100g/mL
Boiling Point 25.0°C (10.0 mmHg)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4060 to 1.4090
Linear Formula C2H5O2CNCO
Quantity 1 g
Specific Gravity 1.11
Formula Weight 115.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Ethoxycarbonyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only