Learn More
Ethoxyacetic acid, 98%
CAS: 627-03-2 | C4H8O3 | 104.11 g/mol
$1487.46 - $1487.46
Chemical Identifiers
| CAS | 627-03-2 |
|---|---|
| Molecular Formula | C4H8O3 |
| Molecular Weight (g/mol) | 104.11 |
| MDL Number | MFCD00004310 |
| InChI Key | YZGQDNOIGFBYKF-UHFFFAOYSA-N |
| Synonym | ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294 |
| PubChem CID | 12301 |
| IUPAC Name | 2-ethoxyacetic acid |
| SMILES | CCOCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC159861000
|
Thermo Scientific Chemicals
159861000 |
100 g | Glass bottle |
Each for $1,487.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 627-03-2 | |
| 104.11 | |
| YZGQDNOIGFBYKF-UHFFFAOYSA-N | |
| 12301 | |
| CCOCC(=O)O |
| C4H8O3 | |
| MFCD00004310 | |
| ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294 | |
| 2-ethoxyacetic acid |
Specifications
| 627-03-2 | |
| 1.1020g/mL | |
| 97°C | |
| 98% | |
| C4H8O3 | |
| C2H5OCH2CO2H | |
| 100 g | |
| 1.102 | |
| Solubility in water: soluble in water | |
| CCOCC(=O)O | |
| 104.11 | |
| 104.11 | |
| Liquid |
| Colorless to Yellow | |
| 97°C to 100°C (11.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4170 to 1.42 | |
| MFCD00004310 | |
| 03, 233 | |
| ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294 | |
| YZGQDNOIGFBYKF-UHFFFAOYSA-N | |
| 2-ethoxyacetic acid | |
| 12301 | |
| 98% | |
| Ethoxyacetic acid, 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 210-978-2
RTECSNumber : AH3600000
TSCA : TSCA
RUO – Research Use Only