missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyacetic acid, 98%

Catalog No. AC159861000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159861000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC159861000 Supplier Thermo Scientific Chemicals Supplier No. 159861000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 627-03-2 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-03-2
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00004310
InChI Key YZGQDNOIGFBYKF-UHFFFAOYSA-N
Synonym ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294
PubChem CID 12301
IUPAC Name 2-ethoxyacetic acid
SMILES CCOCC(=O)O

Specifications

Color Colorless to Yellow
Density 1.1020g/mL
Boiling Point 97°C to 100°C (11.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4170 to 1.42
Linear Formula C2H5OCH2CO2H
Quantity 100 g
Beilstein 03, 233
Specific Gravity 1.102
Solubility Information Solubility in water: soluble in water
Formula Weight 104.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethoxyacetic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 210-978-2
RTECSNumber AH3600000
TSCA TSCA

RUO – Research Use Only