missing translation for 'onlineSavingsMsg'
Learn More

Ethoxy(pentafluoro)cyclotriphosphazene 98.0+%, TCI America™

Catalog No. p-7129356
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11401G 1 g
E11405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11401G Supplier TCI America Supplier No. E11401G
Only null left

Chemical Identifiers

CAS 33027-66-6
Molecular Formula C2H5F5N3OP3
Molecular Weight (g/mol) 275.00
MDL Number MFCD28386107
InChI Key CBTAIOOTRCAMBD-UHFFFAOYSA-N
Synonym 2-Ethoxy-2,4,4,6,6-pentafluoro-1,3,5,2,4,6-triazatriphosphorine
PubChem CID 23419249
IUPAC Name 2-ethoxy-2,4,4,6,6-pentafluoro-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
SMILES CCOP1(F)=NP(F)(F)=NP(F)(F)=N1

Specifications

Color Colorless
Boiling Point 42°C
Quantity 1 g
UN Number 1760
Formula Weight 275.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethoxy(pentafluoro)cyclotriphosphazene
EINECSNumber (5)-6916
TSCA No
Recommended Storage Refrigerator