missing translation for 'onlineSavingsMsg'
Learn More

Ethosuximide

CAS: 77-67-8 | C7H11NO2 | 141.17 g/mol

$326.22 - $326.22

Chemical Identifiers

CAS 77-67-8
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00072123
InChI Key HAPOVYFOVVWLRS-UHFFFAOYNA-N
Synonym ethosuximide, zarontin, etosuximida, 2-ethyl-2-methylsuccinimide, ethosuxide, ethosuccimide, ethosuccinimide, petnidan, suxinutin, atysmal
PubChem CID 3291
ChEBI CHEBI:4887
IUPAC Name 3-ethyl-3-methylpyrrolidine-2,5-dione
SMILES CCC1(C)CC(=O)NC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC461400250
View Documents
Thermo Scientific Chemicals
461400250
25 g
Each for $326.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

77-67-8
141.17
HAPOVYFOVVWLRS-UHFFFAOYNA-N
3291
3-ethyl-3-methylpyrrolidine-2,5-dione
C7H11NO2
MFCD00072123
ethosuximide, zarontin, etosuximida, 2-ethyl-2-methylsuccinimide, ethosuxide, ethosuccimide, ethosuccinimide, petnidan, suxinutin, atysmal
CHEBI:4887
CCC1(C)CC(=O)NC1=O

Specifications

77-67-8
134.0°C (5.0 mmHg)
C7H11NO2
25 g
Solubility in water: soluble.
CCC1(C)CC(=O)NC1=O
141.17
CHEBI:4887
≥97.5% (HPLC)
47.0°C to 50.0°C
Authentic
MFCD00072123
ethosuximide, zarontin, etosuximida, 2-ethyl-2-methylsuccinimide, ethosuxide, ethosuccimide, ethosuccinimide, petnidan, suxinutin, atysmal
HAPOVYFOVVWLRS-UHFFFAOYNA-N
3-ethyl-3-methylpyrrolidine-2,5-dione
3291
141.17
Ethosuximide
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed. Suspected of causing genetic defects. May damage the unborn child.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 201-048-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only