missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethosuximide 98.0+%, TCI America™
$80.16 - $229.29
Chemical Identifiers
| CAS | 77-67-8 |
|---|---|
| Molecular Formula | C7H11NO2 |
| Molecular Weight (g/mol) | 141.17 |
| MDL Number | MFCD00072123 |
| InChI Key | HAPOVYFOVVWLRS-UHFFFAOYNA-N |
| Synonym | ethosuximide, zarontin, etosuximida, 2-ethyl-2-methylsuccinimide, ethosuxide, ethosuccimide, ethosuccinimide, petnidan, suxinutin, atysmal |
| PubChem CID | 3291 |
| ChEBI | CHEBI:4887 |
| IUPAC Name | 3-ethyl-3-methylpyrrolidine-2,5-dione |
| SMILES | CCC1(C)CC(=O)NC1=O |
Chemical Identifiers
| 77-67-8 | |
| 141.17 | |
| HAPOVYFOVVWLRS-UHFFFAOYNA-N | |
| 3291 | |
| 3-ethyl-3-methylpyrrolidine-2,5-dione |
| C7H11NO2 | |
| MFCD00072123 | |
| ethosuximide, zarontin, etosuximida, 2-ethyl-2-methylsuccinimide, ethosuxide, ethosuccimide, ethosuccinimide, petnidan, suxinutin, atysmal | |
| CHEBI:4887 | |
| CCC1(C)CC(=O)NC1=O |
Specifications
| 77-67-8 | |
| White-Yellow | |
| C7H11NO2 | |
| 5 g | |
| HAPOVYFOVVWLRS-UHFFFAOYNA-N | |
| 3-ethyl-3-methylpyrrolidine-2,5-dione | |
| 3291 | |
| 141.17 | |
| Crystalline Powder |
| 51°C | |
| 150°C | |
| MFCD00072123 | |
| ethosuximide, zarontin, etosuximida, 2-ethyl-2-methylsuccinimide, ethosuxide, ethosuccimide, ethosuccinimide, petnidan, suxinutin, atysmal | |
| CCC1(C)CC(=O)NC1=O | |
| 141.17 | |
| CHEBI:4887 | |
| ≥98.0% (GC,T) | |
| Ethosuximide |
Safety and Handling
EINECSNumber : (5)-3375&(5)-0123
RTECSNumber : WN2800000
TSCA : No
Recommended Storage : Refrigerator