missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethosuximide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 51°C |
| Color | White-Yellow |
| Boiling Point | 150°C |
| Quantity | 5 g |
| Formula Weight | 141.17 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethosuximide |
Safety and Handling
| EINECSNumber | (5)-3375&(5)-0123 |
| RTECSNumber | WN2800000 |
| TSCA | No |
| Recommended Storage | Refrigerator |