missing translation for 'onlineSavingsMsg'
Learn More

Ethinylestradiol, 98%, Thermo Scientific Chemicals

Catalog No. p-7077892
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461550010 1 g
AC461550050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461550010 Supplier Thermo Scientific Chemicals Supplier No. 461550010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-63-6
Molecular Formula C20H24O2
Molecular Weight (g/mol) 296.41
MDL Number MFCD00003690
InChI Key BFPYWIDHMRZLRN-SLHNCBLASA-N
Synonym ethinyl estradiol, ethynylestradiol, ethynyl estradiol, ethinylestradiol, ethinyloestradiol, ginestrene, 17-ethinylestradiol, progynon c, ethinoral, eticyclin
PubChem CID 5991
ChEBI CHEBI:4903
IUPAC Name (8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Melting Point 180.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 296.41
Percent Purity 98%
Chemical Name or Material Ethinylestradiol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed. May cause cancer.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 200-342-2

RUO – Research Use Only