missing translation for 'onlineSavingsMsg'
Learn More

Ethidium Bromide, Fisher BioReagents

Catalog No. p-125402
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1021 1 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP1021 Supplier Fisher BioReagents Supplier No. BP102 1
  • Ethidium bromide (EtBr)  commonly used as a nucleic acid stain for gel electrophoresis due to its fluorescence when exposed to UV light
  • Ethidium bromide intercalates double-stranded DNA and RNA
  • Can be used to stain gel after electrophoresis or incorporated into the gel and running buffer 
  • EtBr also acts as inhibitor of RNA and DNA polymerase

Chemical Identifiers

CAS 1239-45-8
Molecular Formula C21H20BrN3
Molecular Weight (g/mol) 394.32
MDL Number MFCD00011724
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
Synonym ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
PubChem CID 14710
ChEBI CHEBI:4883
IUPAC Name 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Melting Point 260°C
Color Purple
pH 4.0 to 7.0
Loss on Drying 5% max.
Packaging Amber Glass
Quantity 1 g
Merck Index 15, 4769
Formula Weight 394.32
Physical Form Powder or Solid
Chemical Name or Material Ethidium Bromide
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Recommended Storage RT