missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine hydrochloride, 98+%

Catalog No. AAL1432222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1432222 100 g
AAL1432236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1432222 Supplier Thermo Scientific Chemicals Supplier No. L1432222

CAS: 2002-24-6 | C2H8ClNO | 97.542 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2002-24-6
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.542
MDL Number MFCD00012892
InChI Key PMUNIMVZCACZBB-UHFFFAOYSA-N
Synonym 2-aminoethanol hydrochloride, ethanolamine hydrochloride, ethanolamine hcl, mea hydrochloride, monoethanolamine hydrochloride, ethanolamine chloride, colamine hydrochloride, ethanol, 2-amino-, hydrochloride, ethanolamine-hcl, 2-hydroxyethylammonium chloride
PubChem CID 74819
IUPAC Name 2-aminoethanol;hydrochloride
SMILES C(CO)N.Cl

Specifications

Melting Point 83°C to 86°C
Quantity 100 g
Beilstein 3542892
Sensitivity Hygroscopic
Merck Index 14,3727
Formula Weight 97.55
Percent Purity ≥98%
Chemical Name or Material Ethanolamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-900-6
RTECSNumber KJ6370000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only