Learn More
Ethanolamine, ACS reagent
CAS: 141-43-5 | C2H7NO | 61.08 g/mol
Supplier: Thermo Scientific Chemicals 451762500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethanolamine | |
| 10.3°C | |
| 170.0°C | |
| Authentic | |
| 1.4530 to 1.4550 | |
| MFCD00008183 | |
| 1.012 | |
| ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
| HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
| 2-aminoethanol | |
| 700 | |
| CHEBI:16000 | |
| ≥99.0% (GC) |
| 141-43-5 | |
| 1.0120g/mL | |
| 92°C | |
| C2H7NO | |
| H2NCH2CH2OH | |
| 250 mL | |
| 15,3782 | |
| Solubility in water: soluble. Other solubilities: miscible with methanol and acetone, soluble in benzene, ether, ccl4 and n-heptane | |
| NCCO | |
| 61.08 | |
| 24 mPa.s (20°C) | |
| 61.08 | |
| ACS Reagent |
Chemical Identifiers
| 141-43-5 | |
| 61.08 | |
| HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
| 700 | |
| NCCO |
| C2H7NO | |
| MFCD00008183 | |
| ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
| CHEBI:16000 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
GHS Signal Word: Danger
EINECSNumber : 205-483-3
RUO – Research Use Only