missing translation for 'onlineSavingsMsg'
Learn More

Ethanol-Benzene Mixture, 1:2, LabChem™

Catalog No. LC141004
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC141004 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC141004 Supplier LabChem Supplier No. LC141004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Ethanol-Benzene Mixture
Name Note 1:2
Color Colorless
CAS Max % 66.67, 30, 1.665, 1.665
CAS 64-17-5, 67-63-0, 67-56-1
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Quantity 4 L
Identification Passes Test
Packaging Amber Glass
Physical Form Liquid
Molecular Formula C6H6
Linear Formula C6H6
MDL Number MFCD00003009
Synonym benzol, cyclohexatriene, benzole, pyrobenzole, benzine, phenyl hydride, pyrobenzol, benzen, phene, mineral naphtha
InChI Key UHOVQNZJYSORNB-UHFFFAOYSA-N
SMILES C1=CC=CC=C1
IUPAC Name benzene
Molecular Weight (g/mol) 78.11
Formula Weight 78.11
Show More Show Less