missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonyl Chloride 98.0+%, TCI America™

Catalog No. E003525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E003525G 25 g
E0035250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E003525G Supplier TCI America Supplier No. E003525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 594-44-5
Molecular Formula C2H5ClO2S
Molecular Weight (g/mol) 128.57
MDL Number MFCD00007460
InChI Key FRYHCSODNHYDPU-UHFFFAOYSA-N
Synonym ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride
PubChem CID 11667
IUPAC Name ethanesulfonyl chloride
SMILES CCS(Cl)(=O)=O

Specifications

Color Yellow
Boiling Point 171°C
Quantity 25 g
UN Number 3265
Formula Weight 128.57
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethanesulfonyl Chloride
EINECSNumber (2)-3915
RTECSNumber KI8050000
TSCA Yes